Learn More
Electron Microscopy Sciences Miltex Biopsy Punch with Plunger 1.5 mm, Yellow
Supplier: Electron Microscopy Sciences 6903102
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Miltex Biopsy Punch with Plunger is a single-use, disposable multi-purpose sampling tool for soft substrates. The plastic exterior is ribbed for comfort and control. The stainless steel cutting tip is seamless and razor sharp to avoid tearing or pulling of the sample. The plunger easily expels the specimen from the barrel.
These sterilized punches come in color-coded blister packs with a 5-year shelf life. In addition to the color-coding, the size is also clearly marked on each punch.
These tools are perfect for tissue processing or forensic applications. For applications where contamination is not an issue, the Miltex Biopsy Punch with Plunger is reusable after cleaning. To clean the cutting edge, rinse with ethanol or spray with compressed air to remove any contaminants.
Color: Yellow
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.