Learn More
MilliporeSigma™ Millistak+™ HC Pod Depth Filter
MilliporeSigma™ Millistak+™ HC Pod Depth Filter features a self-contained and disposable system that protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Supplier: MilliporeSigma™ MD0HC05FS1
Description
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Millistak+™ HC dual-action media improves prefiltration and compresses clarification
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
- Aqueous extraction contains <0.25 EU/mL as determined by the Limulus Amebocyte Lysate (LAL) clot test technique
- 1 autoclave cycle of 60 min at 123°C
Specifications
Cellulose, Inorganic Filter Aid, Glass Fiber Reinforced Polypropylene | |
D0HC | |
Millistak+ HC Pod Depth Filter | |
32 cm |
Primary clarification directly out of the bioreactor | |
3.5 bar (50 psi) at 25°C; 1 bar (15 psi) at 80°C | |
62 cm | |
0.55 m2 |