Learn More
Honeywell™ Miller™ Lined Nylon Body Belt
Specifications
Certifications/Compliance | Meets OSHA, ANSI and CSA Z259.1-95 requirements. |
---|---|
D-Ring Location | One |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-074-573
|
Honeywell™
123NL/LBK |
Large | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
19-074-572
|
Honeywell™
123NL/MBK |
Medium | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
19-074-571
|
Honeywell™
123NL/SBK |
Small | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ideal positioning accessory for full-body harness
Made of durable lined nylon; features a tongue buckle and a single D-ring. Meets OSHA, ANSI and CSA Z259.1-95 requirements. A body belt should be used only for work positioning, or in conjunction with a full-body harness.
Note: OSHA requires that full-body harnesses be used for fall protection in the construction industry.
Specifications
Meets OSHA, ANSI and CSA Z259.1-95 requirements. | |
One |