Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Argos Technologies™ Micryo Label Rolls
Made from chemically inert polyester
Supplier: Argos Technologies™ L1182/EMD
Description
- Unique adhesive formulated to adhere to laboratory glass and plasticware
- Withstand conventional freezing (-80°C) and can also be used for most cryogenic storage applications (vapor and liquid phase nitrogen storage at (-196°C)
- No need for separate labeling solutions for cryo-preservation units or ultra low freezers
- Can even be used in incubators
Specifications
White | |
Polyester | |
0.5 to 2mL tube, can even be used in incubators | |
1000/Roll | |
0.5 in. |
Label | |
Micryo Dot | |
-196°C | |
-320.8°F | |
1.27 cm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction