Learn More
Strategic Applications Inc Microvolume Capillary Blood Collection Tubes, K3EDTA, 20 uL
Supplier: Strategic Applications Inc POCTE20
Ideal for collection of microvolumes of blood via capillary action - from 10ul to 200ul. Small cap aids in transport. Plunger provides quick and clean sample delivery. Available in Lithium Heparin, K3 EDTA or Neutral (untreated). ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.