Learn More
Sarstedt Inc Microvette CB 300, EDTA, 100/Pack
Supplier: Sarstedt Inc 16.444.100
Tube, Capillary; Microvette CB 300; With Push Cap; 300uL; Hematology/Potassium EDTA Preparation; Optimum serum/plasma recovery after centrifugation; Compatible with different analysers for direct sampling