Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MicroGuard MP™ Microporous Coveralls
These Microporous Coveralls incorporate lamination technology, creating a microporous garment with a superior particle and liquid splash barrier.
$126.00 - $156.00
Specifications
Gender | Unisex |
---|---|
Material | Microporous Fabric |
Color | White |
Disposable | Yes |
Waist Style | Elastic |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-472-756
|
MicroGuard MP™
8012S |
Small |
Case of 25 for $126.00
|
|
19-472-755
|
MicroGuard MP™
8012M |
Medium |
Case of 25 for $126.00
|
|
19-472-754
|
MicroGuard MP™
8012L |
Large |
Case of 25 for $126.00
|
|
19-472-757
|
MicroGuard MP™
8012XL |
X-Large |
Case of 25 for $126.00
|
|
19-472-750
|
MicroGuard MP™
80122XL |
2X-Large |
Case of 25 for $126.00
|
|
19-472-751
|
MicroGuard MP™
80123XL |
3X-Large |
Case of 25 for $136.00
|
|
19-472-752
|
MicroGuard MP™
80124XL |
4X-Large |
Case of 25 for $142.00
|
|
19-472-753
|
MicroGuard MP™
80125XL |
5X-Large |
Case of 25 for $156.00
|
|
Description
These Microporous Coveralls are as comfortable as they are safe. Our state-of-the-art lamination technology allows for maximum breathability. A soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so you stay cool even in hot or damp environments. This coverall offers base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.- Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
- Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric.
- Elastic back stretches with user for more comfort and better tear resistance.
- Elastic wrists provide a secure fit and help keep contaminants out.
- Environmentally friendly - can be recycled after use.
Specifications
Unisex | |
White | |
Elastic | |
Elastic | |
None | |
Serged | |
General | |
None |
Microporous Fabric | |
Yes | |
Open | |
Collared | |
Zipper with storm flap | |
Non-sterile | |
Elastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title