Learn More
MicroGuard MP™ Microporous Coveralls
Description
These Microporous Coveralls are as comfortable as they are safe. State-of-the-art lamination technology allows for maximum breathability. A soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so you stay cool even in hot or damp environments. This coverall offers base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection. Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Elastic wrists provide a secure fit and help keep contaminants out
- Environmentally friendly - can be recycled after use
Specifications
Size | Small |
Gender | Unisex |
Material | Microporous Fabric |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Open |
Cuff Style | Elastic |
Neck Style | Collared |
Hood Type | Unhooded |
Show More |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.