Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MicroGuard MP™ Microporous Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
These Microporous Coveralls are as comfortable as they are safe. State-of-the-art lamination technology allows for maximum breathability. A soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so you stay cool even in hot or damp environments. This coverall offers base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection. Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas
- A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
- Elastic back stretches with user for more comfort and better tear resistance
- Elastic wrists provide a secure fit and help keep contaminants out
- Environmentally friendly - can be recycled after use
Specifications
Size | Large |
Gender | Unisex |
Material | Microporous Fabric |
Color | White |
Disposable | Yes |
Waist Style | Elastic |
Ankle Style | Open |
Cuff Style | Elastic |
Neck Style | Collared |
Hood Type | None |
Show More |