Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M Micropore™ Surgical Paper Tape
Micropore™ Surgical Paper Tape - Low-tack adhesive removes gently
Supplier: 3M MMM 1530S1
This item is not returnable.
View return policy
Description
- Latex-free
- Gentle to skin yet adheres well
- Leaves minimal adhesive residue upon removal
Specifications
Paper | |
1.5 yd | |
2.54 cm | |
500/Cs. | |
1530 S-1 | |
General Purpose | |
Acrylate |
Surgical Tape | |
1.37 m | |
1 in. | |
1.5 yd. x 1 in. (9 m x 2.54 cm) | |
No | |
White | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction