Learn More
Cole-Parmer Stuart SI19 Microplate Incubator, 20 L, 120 V
Supplier:  Cole-Parmer 3906510
Encompass_Preferred
Stuart incubator uses forced air circulation to blow preheated air across the shelves for excellent temperature distribution. In conventional incubators, microplates often block the air circulation resulting in uneven temperature in the chamber. A water tray at the bottom of the incubator lets you increase the humidity within the chamber to prevent microplates from drying out. Quickly set and view the temperature on the bright LED display. Easily access the interior with the upwards opening acrylic door. The stainless steel chamber features four shelves each accommodating 6 plates (or 12 stacked in two layers). Shelves slide out for easy loading.
Catalog No. 50-196-0433