Learn More
OHAUS™ Microplate Holder
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Designed to hold standard microplate
Supplier: OHAUS™ 30400213/EMD
Description
- For use with Ohaus Microplate Vortex Mixer
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Two standard microplates | |
Plasitc | |
Re-Suspending Liquids, Mixing Reagents, Buffers and Emulsions, ELISA Assays, Drug Extractions, Mixing Samples, PCR, Digestions, Cell Disruptions, Genotyping | |
1 Ea. |
Double Microplate Holder | |
Double Microplate Holder | |
Microplate Vortex Mixer/Heavy Duty Vortex Mixer |