Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micronova™ PharmaPly™ Mop Head
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Ideal for covering large surface areas quickly and efficiently
Supplier: Micronova™ PPM4MM15
Description
- Polyester/rayon blend mop head for cleaning/disinfecting pharmaceutical support areas, hospitals and laboratories
- Designed for use with MicroMop™ adapter and handle (sold separately)
- 15 in. long (38cm)
Specifications
Cleanroom Mop | |
Non-sterile | |
6 in. | |
Polyester, Rayon |
15 in. | |
Autoclavable | |
Used for Covering Large Surface Areas Quickly and Efficiently, Floor |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction