Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micronova™ AboutFace™ Make-Up Remover
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
A poly-blend cleansing wipe that is pre-moistened with a gentle, alcohol-free cleansing lotion, designed for the face and hands. Wipes are individually packaged and ideal for Pre-Gown Rooms.
Supplier: Micronova™ MMR01
This item is not returnable.
View return policy
Description
The Micronova About Face™ Cleansing Wipes can be used as an effective make-up remover or as a pre-cleaning wipe to remove surface soil prior to gloving or sanitizing hands.
- Individually packaged, poly blend cleansing wipe
- Pre-moistened with a gentle, alcohol-free cleansing lotion
- Highly effective as make-up remover
- Removes surface soil prior to gloving or sanitizing hands
- Ideal for Pre-Gown Rooms
- Biodegradable
Specifications
Pre-saturated Face Wipe | |
7.6 cm | |
7.6 cm | |
Not Autoclavable | |
Store at ambient temperature | |
Biodegradable Synthetic Wipe | |
250/Cs. |
3 in. | |
3 in. | |
Poly Blend | |
White | |
Single-use | |
For Make up Removal, Remove Surface Grime, Pre-Moistened Cleansing Wipe | |
Non-sterile |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction