Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeves
Micromesh™ Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeves are molded from a special high density polymer to keep specimens safely submerged in liquid.
$334.00 - $344.00
Specifications
Quantity 75 sleeves/Cs.
Writing Face Angle 45°
Material Acetal
View More Specs
Products 11
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
23-045-363
View Documents
Simport™ Scientific
M50710SL
Lilac
Case of 750 for $344.00
Add to Cart
23-045-364
View Documents
Simport™ Scientific
M50711SL
Orange
Case of 750 for $344.00
Add to Cart
23-045-365
View Documents
Simport™ Scientific
M50712SL
Aqua
Case of 750 for $334.00
Add to Cart
23-045-366
View Documents
Simport™ Scientific
M5072SL
White
Case of 750 for $344.00
Add to Cart
23-045-367
View Documents
Simport™ Scientific
M5073SL
Pink
Case of 750 for $334.00
Add to Cart
23-045-368
View Documents
Simport™ Scientific
M5074SL
Green
Case of 750 for $344.00
Add to Cart
23-045-369
View Documents
Simport™ Scientific
M5075SL
Yellow
Case of 750 for $344.00
Add to Cart
23-045-370
View Documents
Simport™ Scientific
M5076SL
Blue
Case of 750 for $334.00
Add to Cart
23-045-371
View Documents
Simport™ Scientific
M5077SL
Peach
Case of 750 for $334.00
Add to Cart
23-045-372
View Documents
Simport™ Scientific
M5078SL
Tan
Case of 750 for $334.00
Add to Cart
23-045-373
View Documents
Simport™ Scientific
M5079SL
Gray
Case of 750 for $334.00
Add to Cart
Description
Description
  • Totally resistant to the chemical action of histological solvents.
  • Lid can be opened and closed as often as necessary and always re-locks securely without danger of specimen loss.
  • Offers 1,676 square openings (0.38 mm) allowing for a greatly improved fluid exchange and drainage without having to use biopsy pads.
  • Large anterior slots in both cassette and cover ensure that the Micromesh™ biopsy cassette will sink rapidly.
  • Large square compartment measuring 27 mm is ideal even for needle biopsies.
  • Cover does not protrude above the cassette, an excellent space-saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
  • Secure locking device safely holds the specimen during processing.
  • Convenient Quickload™ sleeves are suitable for automated printers.
Specifications
Specifications
75 sleeves/Cs.
45°
Acetal