Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with FEP/Natural Rubber Septa
Available in multiple colors and package sizes.
Supplier: DWK Life Sciences 090020NB
Specifications
Natural | |
No | |
2000 | |
Screw Cap | |
9 mm Autosampler Vials - MicroLiter |
Polypropylene | |
FEP, Natural Rubber, FEP | |
MicroLiter | |
9 mm |