Learn More
DWK Life Sciences MicroLiter 76 Series™ 9 mm Autosampler Vial, Cap and Septa Unassembled Component Kits with PTFE/Silicone/PTFE Septa
SDP
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Supplier:  DWK Life Sciences 761040B
 View more versions of this product
Catalog No. 15-111-847
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Blue
12 mm
No
PTFE, Silicone, PTFE
MicroLiter
Screw Cap
9 mm
Clear
Borosilicate Glass (Type 1)
2 mL
100
Polypropylene
32 mm
No
Product Suggestions
Product Suggestions