Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences MicroLiter 11mm AlumiTin Magnetic Crimp Top Autosampler Vial Seals
MicroLiter's 11mm AlumiTin crimp-top autosampler vial seals are available in multiple colors and septa types.
Supplier: DWK Life Sciences 110030ATB
Specifications
PTFE/Silicone | |
Aluminum | |
PTFE/Silicone | |
Crimp Cap | |
11 mm Autosampler Vials - MicroLiter |
Silver | |
No | |
2000 | |
11 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction