Learn More
World Precision Instrument Microinjector with External Pressure Source (pressure source not included)
SDP
Supplier:  World Precision Instrument PV850
Encompass_Preferred
For All Your Microinjection Needs  
WPI's new generation pressure microinjectors (the PV850, µPUMP, and MICRO-ePUMP) are quickly replacing the popular legacy line of pressure injectors (the PV820 and PV830). Our three new injectors facilitate any type of microinjecion that involves glass micropipettes, conveniently paired with a compensation pressure function to prevent capillary action. 
Now featuring: Narishige & TransferMan-compatible capillary holder included with each unit! 
To learn more about our warranty options, .
Catalog No. 50-253-8619