Learn More
Perkin Elmer US LLC Microfocus Beam Condenser - KBr
Supplier:  Perkin Elmer US LLC L1272284
Encompass_Preferred
The Microfocus Beam Condenser is a simple two lens linear optical system that ensures easy alignment and high throughput over a wide working wavelength range. The required DC-2 Diamond Compression Cell compresses samples to a thickness that is ideal for transmission experiments. .
Catalog No. 50-211-3513