Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell MICROFLEX™ Lifestar EC™ LSE-104 High Risk Nitrile Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Designed specifically for EMS and healthcare professionals
- Durable for safe critical patient care
- Dual layer, dual color design for two layers of protection
- Bright blue interior makes breaches more detectable
- White exterior makes fluid and blood contact more visible
- Tested against both fentanyl and gastric acid to simulate hazardous, real world overdose situations
- Advanced barrier protection (AQL 0.65)
- Non-stick and non-foaming properties for reduced interference when working
Specifications
Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600, ISO 13485, ISO 9001, NFPA 1999:2013 |
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
Chemotherapeutic Rated | No |
Applications | Healthcare |
AQL | 0.65 |
Color | White, Blue |
Disposable | Single-use |
Exterior Finish | Partially Textured |
Finger Thickness (English) | 8.3 mil |
Finger Thickness (Metric) | 0.21 mm |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction