Learn More
World Precision Instrument Microelectrode Holder (MPH6R)-MPH6R10
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument MPH6R10
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For use with these WPI Products: Piconozzle Kit (5430-XX)
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.