Learn More
Fine Science Tools Burrs for Micro Drill, Carbon Steel, 0.9 mm Tip Dia, 44 mm, 10/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1900709
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tip Diameter: 0.9mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No