Learn More
Carson MicroBrite Pro Microscope
Carson MicroBrite Pro 60x-120x LED Lighted Zoom Pocket Microscope with Aspheric Lens System and Smartphone Digiscoping Clip (MM-350).
Supplier: Carson MM350
This item is not returnable.
View return policy
Description
- The Carson MicroBrite Pro LED Lighted Pocket Microscope is designed to be lightweight and compact without sacrificing performance.
- It has precision molded aspheric lenses that provide high-quality optics with 60x-120x magnification and clear vision.
- The MicroBrite Pro Smartphone Adapter Clip allows you to easily fit the microscope onto most smartphones, so you can share images and videos with others.
- The microscope is suitable for students, professionals, and anyone interested in observing the natural world.
- It's ideal for appraising jewelry, currency, or stamps, as well as observing microorganisms, coins, food, fabric, and trichomes.
Specifications
Monocular | |
Clip-on | |
1.5 V | |
Single Precision Molded Aspheric Lens |
Microscope | |
LED | |
60X to 120X |