Learn More
Microbiologics™ Microbiology QC Slides
Microscope slide preparations with one or two droplets of an air-dried and methanol-fixed organism, or a single smear containing a designated organism population
Supplier: Microbiologics™ SL4110
Description
Support formal quality assurance programs by serving as quality control challenges to verify performance of stain reagents, staining methods and the performance of personnel interpreting microscopic examinations.
Acid Fast Control SlideProvides two air-dried and methanol-fixed droplets within two etched circles
- Provides a single, air-dried and methanol-fixed fecal smear containing Acid Fast-Positive Cryptosporidium and acid fast-negative intestinal bacteria
- Provides a single, air-dried and methanol-fixed preparation containing Candida albicans, a derivative of ATCC™ 10231™†
- Provides a single, air dried, methanol-fixed, preparation containing Candida albicans, a derivative of ATCC™ 10231™† with leucocytes and erythrocytes
- Provides two air-dried and methanol-fixed droplets within two etched circles
- Circle nearest the label contains an acid-fast positive Mycobacterium gordonae, a derivative of ATCC™ 14470™†
- Circle farthest contains an acid fast-negative Erysipelothrix rhusiopathiae, a derivative of ATCC™ 19414™†
- Pneumocystis carinii Control Slide
- Provides a single, air-dried and methanol-fixed rat lung tissue preparation containing Pneumocystis
- SAF-Preserved Fecal Smear Control
- Provides an air-dried, SAF preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia
- S. aureus and E. coli Control Slide
- Provides two air-dried and methanol-fixed droplets within two etched circles
- Circle nearest the label contains a droplet of a Gram-Positive Staphylococcus aureus derivative of ATCC™ 25923™†
- Circle furthest from the label contains a droplet of a Gram-negative Escherichia coli derivative of ATCC™ 25922™†
- Provides an air-dried, Zinc PVA-preserved, fecal smear containing a representative intestinal protozoa, usually Giardia lamblia
Specifications
CE | |
Slide | |
Microscopic examination | |
10 Slides |
Control Slide | |
Control Slide | |
Acid fast positive Mycobacterium gordonae ATCC 14470, Acid fast negative Erysipelothrix rhusiopathiae ATCC 19414 |