Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Epower™ Pseudomonas aeruginosa, ATCC™ 27853™†
Lyophilized pellets add convenience to special projects and internal proficiency plans
Specifications
Species | Pseudomonas aeruginosa |
---|---|
ATCC® | 27853 |
Biosafety Level | 2 |
Form | Lyophilized Pellet |
Product Type | QC Microorganism |
Includes: Product warranty and peel-off certificate of assay
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
23-003-5038
|
Microbiologics™
0353E6 |
27853 | Pseudomonas aeruginosa | 1.0-9.9E+06 CFU per pellet |
Pack of 10 for $709.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-820
|
Microbiologics™
0353E3 |
27853 | Pseudomonas aeruginosa | 1.0-9.9E+03 CFU per pellet |
Pack of 10 for $709.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Pseudomonas aeruginosa | |
2 | |
QC Microorganism | |
Product warranty and peel-off certificate of assay | |
10/Pk. |
27853 | |
Lyophilized Pellet | |
Lyophilized pellets add convenience to special projects and internal proficiency plans. | |
Vial |
Safety and Handling
ShelfLife : 10 to 20 Months
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title