Learn More
Microbiologics™ Epower™ Aspergillus brasiliensis ATCC™ 16404™†
Lyophilized pellets add convenience to special projects and internal proficiency plans
Specifications
Species | Aspergillus brasiliensis |
---|---|
ATCC® | 16404 |
Biosafety Level | 1 |
Form | Lyophilized Pellet |
Product Type | QC Microorganism |
Includes: Product warranty and peel-off certificate of assay
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
23-003-3397
|
Microbiologics™
0392E6 |
16404 | Aspergillus brasiliensis | 1.0-9.9E+06 CFU per pellet |
Pack of 10 for $693.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-003-5012
|
Microbiologics™
0392E3 |
16404 | Aspergillus brasiliensis | 1.0-9.9E+03 CFU per pellet |
Pack of 10 for $693.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Microorganism preparations manufactured at a predetermined concentration for versatile laboratory testing requirements.
- Process controls from one hundred to over a billion CFU/preparation
- Shelf life of 10-20 months Traceable to reference cultures
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Aspergillus brasiliensis | |
1 | |
QC Microorganism | |
Product warranty and peel-off certificate of assay | |
10/Pk. |
16404 | |
Lyophilized Pellet | |
Lyophilized pellets add convenience to special projects and internal proficiency plans. | |
Vial |
Safety and Handling
ShelfLife : 10 to 20 Months
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.