Learn More
SPEX CERTIPREP LLC 3P Microbe Cocktail with Yeast and Mold, 10 Sample Set
Supplier: SPEX CERTIPREP LLC 3P FM73310
(NSI by Zeptometrix) This multiple organism blended CRM is designed for use with Petrifilm, other re-hydratable film methods or BAM methods. CRM is quantitatively certified for APC, Total coliforms, E. coli, Enterobacteriaceae, S. aureus, Mold and Yeast within the method countable ranges after hydration.