Learn More
Kamiya Biomedical Microalbumin Calibrator, 6x1mL/PK RX
Supplier: Kamiya Biomedical KAI020C
K-ASSAY Microalbumin Calibrator. 6 x 1 mL (liquid). For 6 point calibration. The K-ASSAY Microalbumin Calibrator is for the calibration of the K-ASSAY Microalbumin assay (Cat. No. KAI-019 or KAI-057) for quantifying albumin in urine specimens. FOR IN VITRO DIAGNOSTIC USE.