Learn More
World Precision Instrument Micro Ophthalmic Sterilization Baskets with Lids-WP-22001
Supplier: World Precision Instrument WP9022001
- Designed specifically for delicate micro instruments
- Silicon inserts on bottom baskets to accommodate micro instruments
- Lids are provided with silicon strips to grip the instruments
- Side walls are made of perforated sheets
- User friendly locking handle
- Manufactured with stainless steel 304
- Electrolyte polished