Learn More
Electron Microscopy Sciences Micro Friedman Bone Rongeur
Supplier: Electron Microscopy Sciences 7798101
Bone Rongeur, curved. Very delicate. The jaw width is 1.3 mm. and it is 5 1/2" long. Made from stainless steel with a polished finish.