Learn More
Fine Science Tools Trephines for Micro Drill, 2.7mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1800427
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tip Diameter: 2.7mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Stainless Steel/ Length: 44mm/ Autoclave Safe: Yes. These trephines with a side opening are ideal for clearing bone and other materials.