Learn More
Roboz Surgical MICRO CLIP SLT CRV 1X6MM
Supplier: Roboz Surgical RS5431
Micro Clip; Slight Curve; 70 Grams Pressure; 1mm Clip Width; 6mm Jaw Length The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and