Learn More
Cole-Parmer Cole-Parmer Micro-90 cleaning solution, 12/cs
Supplier: Cole-Parmer 1810004
Micro-90 is a mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications. Solution cleans glass, most metals, plastic, rubber, fabric, and is safe for all your ultrasonic cleaners. Radioactive contaminants will not reduce cleaning capability. Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water. Solution is environmentally friendly—contains no phosphates, CFCs, ODCs, or borates. It safely replaces chromic acid to remove protein from glassware. Designed for use in CIP, ultrasonic and immersion applications. Can be used in validated processses. A 1% to 2% solution is effective in most applications. Typical pH: 9.5. NSF-Registered for USDA A1 use. Note: Do not use on zinc, magnesium, or aluminum. 18100-30 shipped FOB Burlington, NJ, USA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.