Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Mibefradil dihydrochloride
CaV3.x blocker
Specifications
Purity | >97% |
---|---|
Product Type | Mibefradil dihydrochloride |
Molecular Weight (g/mol) | 568.56 |
Inhibitors | Mibefradil dihydrochloride |
Description
Ca2+ channel blocker with moderate selectivity for T-type Ca2+ channels displaying IC50 values of 2.7 μM and 18.6 μM for T-type and L-type channels respectively. Antihypertensive agent.Specifications
>97% | |
Mibefradil dihydrochloride | |
568.56 | |
Mibefradil dihydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title