Learn More
AdipoGen MgATP Solution
SDP
Supplier:  AdipoGen AG10T0022R100
Encompass_Preferred
Chemical. Synthetic. Pre-coupled Mg-ATP is an ideal energy source for semi-purified conjugation/degradation reactions. Supplied as a pH adjusted solution and will not alter reaction pH. Eliminates the reaction needed to produce usable ATP when MgCl2 and ATP are added separately. The amount provided is sufficient for 10 to 50 reactions depending on conditions.
Catalog No. 50-256-3399
Explore available promotions