Learn More
Epigentek Inc MethylFlash Methylated DNA 5-mC Quantification Kit (Colorimetric) (48 assays)
Supplier: Epigentek Inc P103448
For absorbance-based quantitation of global DNA methylation in an ELISA-like format