Learn More
Mettler Toledo™ Connection Tube for Mettler Toledo Karl Fischer™ Titrators
Accessory for use with Mettler Toledo Karl Fischer Titrators.
Supplier: Mettler Toledo™ 23957
Description
- Connects titration head to titration beaker drying tube
- For model V20/30 Compact Karl Fischer titrator and DL31/38 Karl Fischer titrator
Specifications
Connection Tube |