Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metric Weight Set
Stackable, color-coded weights come in a plastic storage box.
Supplier: Learning Resources, Inc 32071
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Description
- Twenty each of 1 and 5g weights
- Ten 10g weights
- Four 20g weights
- Two 100g weights
- One each of 500 and 1000g weights
Order Info
Grades 3 to 8.
Specifications
Metric Weight Set | |
Weight: 20 x 1g, 20 x 5g, 10 x 10g, 4 x 20g, 2 x 100g, 500g, 1000g, Plastic storage box |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction