Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metoclopramide Hydrochloride, Monohydrate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog Number 18604552
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
54143-57-6
C14H25Cl2N3O3
354.27
MFCD00150634
KJBLQGHJOCAOJP-UHFFFAOYSA-N
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
54143-57-6
C14H25Cl2N3O3
354.27
MFCD00150634
KJBLQGHJOCAOJP-UHFFFAOYSA-N
4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

ME176, 54143-57-6, C14H22ClN3O2•HCI•H2O

Spectrum™ Chemical Metoclopramide Hydrochloride, Monohydrate, USP is a medication used mostly for stomach and esophageal problems to treat and prevent nausea and vomiting. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 101%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit