Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Methylene Blue Solution, 1%
$6.60 - $14.50
Specifications
Chemical Name or Material | Methylene Blue Solution, 1% |
---|---|
CAS | 7220-79-3, 7732-18-5 |
Assay Percent Range | 1% w/v |
Linear Formula | C16H18ClN3S·3H2O |
Synonym | Methylene Blue Chloride, Basic Blue 9 |
Description
Biological stainSpecifications
Methylene Blue Solution, 1% | |
1% w/v | |
Methylene Blue Chloride, Basic Blue 9 | |
1% | |
Blue |
7220-79-3, 7732-18-5 | |
C16H18ClN3S·3H2O | |
373.9 | |
∽1.00g/mL at 20°C |
Safety and Handling
ShelfLife : 18 months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title