Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methotrexate, USP and EP Grade, MP Biomedicals™
Click to view available options
Quantity:
1 mg
10 mg
25 mg
100 mg
500 mg
1 g
Description
Chemotherapy
Specifications
Color | Yellow |
Quantity | 10 mg |
Formula Weight | 454.46 |
Grade | EP/USP |
Physical Form | Solid |
Chemical Name or Material | Methotrexate |
Safety and Handling
storageNote1 | Protect from light. |
Recommended Storage | Store at 4°C |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction