Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Methionine/Choline Deficient Diet (Powdered)
Methionine/Choline Deficient Diet (Powdered) has been used to study nonalcoholic steatohepatitis (NASH).
Supplier: MP Biomedicals™ 0296043820
Description
- Contains sucrose, corn starch, corn oil, Alphacel non-nutritive bulk, and AIN 76 mineral mix.
Specifications
Powder | |
20 kg |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction