Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol (Pesticide), Fisher Chemical
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™ Amber Glass
Specifications
Melting Point | -98°C |
Color | Colorless |
Linear Formula | CH3OH |
UN Number | 1230 |
Formula Weight | 32.04 |
Grade | Pesticide |
Density | 0.791g/cm³ |
Boiling Point | 64.7°C |
Quantity | 4 L |
Flash Point | 12°C (53.6°F) |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. NFPA Health:1 Flammability:3 Instability:0 |
DOTInformation | Class 3: Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction