Learn More
Cole-Parmer TSI 7545 Advanced IAQ Meter; Datalogging, CO and CO2, with Humidity
Supplier: Cole-Parmer 8638415
Easy-to-use keypad scrolls through all readings at the touch of a button. Low-drift NDIR CO2 sensor gives stable, accurate readings. Check building IAQ conditions to optimize productivity. Ideal for testing HVAC system performance.