Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Metalguard™ 1880 Heavy Weight Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Reinforced thumb crotch for longer life
- Continuous knit for close secure fit
- Ambidextrous
- Prewashed and preshrunk
- ANSI Level A6 cut resistance
- Made in the USA
Specifications
ANSI Abrasion Rating | ANSI 3 |
Certifications/Compliance | ASTM F2992-15, ASTM D3884-09 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | Green, Yellow |
Cut Resistance Rating | ANSI A6 |
Hand | Ambidextrous |
Material | Aramid Fiber, Stainless Steel Fiber |
Product Type | Cut-resistant Gloves |
Size | X-Small |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction