Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Metal Activity Table
SCI_ED

Fixed legs with 1 x 2 in. apron frame.

Supplier:  Diversified Spaces™ X7244

 View more versions of this product

Catalog No. S42486


Add to Cart

Description

Description

  • Fixed legs
  • Black color
  • 1.25 in. square legs
Specifications

Specifications

Metal Activity Table
Fixed legs
Phenolic
36 x 72 x 30 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit