Learn More
Enzo Life Sciences Met enkephalin polyclonal antibody (25µl)
Supplier: Enzo Life Sciences BMLEA11500025
Immunogen: Synthetic methionine enkephalin. Source: Whole rabbit serum. Species reactivity: Species independent. Specificity: Recognizes methionine enkephalin in a wide range of species. Applications: IHC. Formulation: Liquid. Antiserum containing 0.1% sodium azide.