Learn More
Sklar Instruments Forceps, Merit Iris Dressing Forceps, Sklar 4 in., Blunt, Slightly Curved, Serrated, Thumb, Stainless Steel, EA.
Supplier: Sklar Instruments 98135
Forceps, Merit Iris Dressing Forceps, Sklar, 4 in., Blunt, Slightly Curved, Serrated, Thumb, Stainless Steel, EA. Iris Dressing Forceps are thumb style forceps commonly used for placing or removing dressings in many types of procedures. Although Dressing Forceps were originally designed to specifically hold materials such as gauze, they have come to have a variety of additional functions. They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing. Serrated jaws assist in a having a secure hold on items. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.