Learn More
Meridian Bioscience™ Premier™ EHEC Microplate Assay
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Detects all toxin-producing EHEC strains unlike other methods which are limited to E. coli 0157:H7 detection
Supplier: Meridian Bioscience™ 608096
This item is not returnable.
View return policy
Description
- CLIA Complexity: High
- A breakaway microwell EIA for the detection of toxins produced by Enterohemorrhagic E. coli
- Results in just over two hours
- Sample from fresh, frozen, or Cary-Blair preserved stool specimens, broth cultures, or colony sweeps of agar plates
- Simple, easy-to-use procedure contains color-coded, ready-to-use reagents
- Results can be read visually or spectrophotometrically with a standard microplate reader
Specifications
High | |
Premier™ EHEC Test | |
2 hr. |
Shiga Toxins I and II | |
96 Tests |